missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KD Scientific Stainless-Steel Syringes
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | No Needle Included |
| Diameter (English) Inner | 1-37/100 in. |
| Diameter (English) Outer | 1-1/5 in. |
| Diameter (Metric) Inner | 34.7 mm |
| Diameter (Metric) Outer | 38.1 mm |
| Product Type | Luer-Lok Syringe |
| Max. Pressure | 1500 psi |
| Volume (Metric) | 100 mL |
| Show More |