missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNQ5 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PA1941
Modifier l'affichage
Click to view available options
Quantité:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PA1941 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PA1941 Fournisseur Invitrogen™ Code fournisseur PA1941
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

PA1-941 detects the KCNQ5 from rat and human samples. This antibody is specific for KCNQ5 and does not detect KCNQ1, KCNQ2, KCNQ3 or KCNQ4. PA1-941 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~125 kDa band representing KCNQ5 from rat brain extract. The PA1-941 immunizing peptide corresponds to amino acid residues 880-897 from human KCNQ5.

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system. Studies have shown that KCNQ3 and KCNQ5 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy. Voltage-dependent potassium channels are key regulators of the resting membrane potential and modulate the excitability of electrically active cells. The channels are usually tetrameric and can interact with auxiliary subunits that enhance or modify currents mediated by the pore-forming subunits.

Spécifications

Antigène KCNQ5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1mg/mL BSA and 0.05% sodium azide
Gène KCNQ5
Numéro d’acquisition du gène Q9NR82
Alias du gène 7730402H11; 9230107O05Rik; AA589396; D1Mgi1; KCNQ5; Kcnq5l; KQT-like 5; Kv7.5; KvLQT5; potassium channel protein; potassium channel subunit alpha KvLQT5; potassium channel, voltage gated KQT-like subfamily Q, member 5; potassium channel, voltage-gated KQT-like subfamily Q, member 5; potassium voltage-gated channel subfamily KQT member 5; potassium voltage-gated channel subfamily Q member 5; potassium voltage-gated channel, KQT-like subfamily, member 5; potassium voltage-gated channel, subfamily Q, member 5; rCG_43583; voltage-gated potassium channel subunit Kv7.5; voltage-gated potassium channel type Kv7.5
Symboles de gène KCNQ5
Espèces hôtes Rabbit
Immunogène Synthetic peptide corresponding to residues C(880) K A G E S T D A L S L P H V K L K(897) of human KCNQ5 protein.
Méthode de purification Antigen affinity chromatography
Quantité 200 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 259273, 56479
Espèces cibles Human, Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats