missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappler™ Zytron™ 200 Hooded Coveralls
Provide increased protection with resistance to a wider range of chemicals
$591.66 - $1011.88
Specifications
| Material | Zytron™ |
|---|---|
| Color | Gray |
| Neck Style | Hooded |
| Resistance | Chemicals |
| For Use With (Application) | Ideally used with wider range of chemicals |
| Catalog Number | Mfr. No. | Description | Seam Type | Size | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Seam Type | Size | Price | Quantity | |||
|
19-171-140A
|
Kappler™
Z2H414SMMD |
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure | Sealed, Taped | Medium |
Case of 6 Each for $761.17
|
|
|||
|
19-171-140B
|
Kappler™
Z2H414LGXL |
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure | Sealed, Taped | Large |
Case of 6 Each for $761.17
|
|
|||
|
19-171-140D
|
Kappler™
Z2H4142X3X |
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure | Sealed, Taped | 2X-Large |
Case of 6 Each for $837.28
|
|
|||
|
19-171-141C
|
Kappler™
Z2H426LGXL |
Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper | Sealed, Taped | Large, X-Large |
Case of 6 Each for $877.49
|
|
|||
|
19-171-140E
|
Kappler™
Z2H4143X |
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure | Sealed, Taped | 3X-Large |
Case of 6 Each for $837.28
|
|
|||
|
19-171-141A
|
Kappler™
Z2H426SMMD |
Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper | Sealed, Taped | Small, Medium |
Case of 6 Each for $877.49
|
|
|||
|
19-171-141E
|
Kappler™
Z2H4262X3X |
Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper | Sealed, Taped | 2X-Large, 3X-Large |
Case of 6 Each for $1,011.88
|
|
|||
|
19-171-142B
|
Kappler™
Z2B428LGXL |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Bound | Large |
Case of 12 Each for $600.66
|
|
|||
|
19-171-142A
|
Kappler™
Z2B428SMMD |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Bound | Medium |
Case of 12 Each for $600.66
|
|
|||
|
19-171-143D
|
Kappler™
Z2H4282X3XDUP |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Sealed, Taped | 2X-Large |
Case of 6 Each for $650.82
|
|
|||
|
19-171-142D
|
Kappler™
Z2B4282X3X |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Bound | 2X-Large |
Case of 12 Each for $660.82
|
|
|||
|
19-171-143A
|
Kappler™
Z2H428SMMD |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Sealed, Taped | Medium |
Case of 6 Each for $591.66
|
|
|||
|
19-171-143C
|
Kappler™
Z2H428LGXLDUP |
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening | Sealed, Taped | X-Large |
Case of 6 Each for $591.66
|
|
|||
Description
- Attached hood
- Zippered front
- Color: Gray
Applications:
Valuable in applications where rip-out is a concern
Specifications
| Zytron™ | |
| Hooded | |
| Ideally used with wider range of chemicals |
| Gray | |
| Chemicals |