missing translation for 'onlineSavingsMsg'
Learn More

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Supplier:  Kappler™ Z2H426SMMD

 View more versions of this product

Catalog No. 19-171-141A


Only null left
Explore available promotions

Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

Small, Medium
Gray
Elastic
Elastic
Sealed, Taped
CE Type 3
Ideally used with wider range of chemicals
4.1 kg
Zytron™
Attached Socks, Boot Overflaps
Hooded
Double Storm Flap with Hook and Loop Seal
Chemicals
Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper
9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions