missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Powder, USP, Spectrum™ Chemical

Catalog No. 18603624
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603624 2.5 kg Poly Bottle
18603622 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603624 Supplier Spectrum Chemical Mfg Cor Supplier No. KA1052.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

KA105, 1332-58-7

Spectrum™ Chemical Kaolin, Powder, USP is used to treat diarrhea and to relieve soreness and swelling inside the mouth caused by radiation treatments. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Grade USP
Loss on Ignition 15%
Packaging Poly Bottle
Chemical Name or Material Kaolin, Powder