missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kanamycin Monosulfate 94.0+%, TCI America™
$129.10 - $427.20
Chemical Identifiers
| CAS | 25389-94-0 |
|---|---|
| Molecular Formula | C18H38N4O15S |
| Molecular Weight (g/mol) | 582.575 |
| MDL Number | MFCD00070253 |
| InChI Key | OOYGSFOGFJDDHP-IZQIRFRQSA-N |
| Synonym | O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4)]-2-deoxy-D-streptamine Monosulfate |
| PubChem CID | 134129479 |
| IUPAC Name | (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
| SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
Chemical Identifiers
| 25389-94-0 | |
| 582.575 | |
| OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
| 134129479 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
| C18H38N4O15S | |
| MFCD00070253 | |
| O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4)]-2-deoxy-D-streptamine Monosulfate | |
| (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
Specifications
| Kanamycin Monosulfate | |
| 25389-94-0 | |
| White/Yellow | |
| C18H38N4O15S | |
| O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4)]-2-deoxy-D-streptamine Monosulfate | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O | |
| 582.575 | |
| 582.58 | |
| Kanamycin Monosulfate |
| Crystalline Powder | |
| 250°C | |
| 5 g | |
| MFCD00070253 | |
| OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
| (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid | |
| 134129479 | |
| ≥94.0% (N) |
Safety and Handling
RTECSNumber : NZ3225030
TSCA : No