missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Monosulfate 94.0+%, TCI America™

Supplier:  TCI America K00475G

 View more versions of this product

Catalog No. K00475G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Kanamycin Monosulfate
Crystalline Powder
250°C
5 g
MFCD00070253
OOYGSFOGFJDDHP-IZQIRFRQSA-N
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
134129479
≥94.0% (N)
Kanamycin Monosulfate
25389-94-0
White/Yellow
C18H38N4O15S
O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4)]-2-deoxy-D-streptamine Monosulfate
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
582.575
582.58

Chemical Identifiers

25389-94-0
582.575
OOYGSFOGFJDDHP-IZQIRFRQSA-N
134129479
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
C18H38N4O15S
MFCD00070253
O-3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-[6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4)]-2-deoxy-D-streptamine Monosulfate
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NZ3225030

TSCA : No

SDS
Documents

Documents

Promotions

Promotions