missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Piggyback Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Catalog No. 19414806
Change view
Click to view available options
Dimensions (D x W x H) Interior:
14.5 x 39.5 x 13 in.
14.5 x 39.5 x 19.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior
19414806 14.5 x 39.5 x 19.5 in.
19414805 14.5 x 39.5 x 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19414806 Supplier Justrite™ Supplier No. 861728

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Add another level of security to your work environment. The Justrite Sure-Grip™ EX Piggyback Hazardous Material Steel Safety Cabinet segregates hazardous materials using cabinets with an exclusive label package that ensures full regulatory compliance.

  • Designed to mount on top of standard 114L and 170L safety cabinets
  • Used independently as wall-mount unit or on countertop
  • Features double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet
  • Automatic self-close door(s) shut and latch when fusible link melts at 74°C (165°F) under fire conditions
  • Polyethylene trays to mount on top of galvanized steel shelves
  • Separated polyethylene liner for bottom sump resists aggressive chemicals
  • Epoxy coated baked on powder-coat finish on both interior and exterior surface ensures chemical resistance
  • Each cabinet includes Hazardous - Keep Fire Away label and pack of ten application-specific labels to indicate its contents

Compliance

NFPA, U (NFPA)

Warranty

10 Year

Specifications

Dimensions (D x W x H) Interior 14.5 x 39.5 x 19.5 in.
Product Type Combi Safety Storage Cabinet
Door Style Self-close
Load Bearing Capacity 17 gal. (64 L)
No. of Doors 2
No. of Shelves 1
Color Blue
For Use With (Application) Hazardous materials