Learn More
Justrite™ HPLC Manifold Kit for Quick-Disconnect Disposal Cans: 1/16 in. O.D. Tube Fittings
Catalog No. 19160370
Change view
Click to view available options
Diameter (English) Outer:
0.062 in.
0.125 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer
19160370 0.125 in.
19160369 0.062 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19160370 Supplier Justrite™ Supplier No. 28175
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Economical way to safely collect waste from multiple HPLC machines
Use with FM-approved Justrite™ Disposal Cans with stainless-steel quick-disconnect fittings to feed up to four waste lines from HPLC processes into a single intake fitting.
  • Peek fittings include four each: 1/16 in. (1.6mm) and 1/8 in. (3.2mm) O.D. tube fittings with polychlorotrifluoroethylene (PCTFE) ferrules for compression fit of waste line
  • Multiple manifolds can be used on custom-designed disposal cans with stainless-steel fittings (one fitting must remain unused to permit venting and prevent back pressure)
Certifications
FM approved.
Compliance
Safety can (sold separately) with installed manifold meets OSHA and NFPA Code 30 requirements.
Specifications
Certifications/Compliance FM, OSHA, NFPA Code 30
Diameter (Metric) Outer 0.3 cm
Diameter (English) Outer 0.125 in.
Quantity 1 Ea.
Includes ETFE ferrules
For Use With (Application) Safely collect waste from multiple HPLC machines
For Use With (Equipment) Justrite™ disposal cans with stainless steel quick-disconnect fittings
Material Polypropylene
Product Type Tube Fitting