missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Biohazard Waste Can

Catalog No. 19033755
Change view
Click to view available options
Capacity (English):
6 gal.
10 gal.
Diameter (English) Outer:
12 in.
14 in.
Height (English):
16 in.
18 in.
Includes:
Foot-operated Cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Diameter (English) Outer Height (English) Includes
19033755 10 gal. 14 in. 18 in. Foot-operated Cover
19033725 6 gal. 12 in. 16 in. Foot-operated Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19033755 Supplier Justrite™ Supplier No. JMC05930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.

Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.

  • Features leakproof, sturdy steel construction with foot-operated, self-closing lid
  • Eliminates any reason to reach into the waste area or touch the can
  • Holds biohazard bag (sold separately) securely around circumference of can top
  • Large biohazard label clearly identifies the contents and alerts workers

Not to be used for disposal of sharp items.

Certifications

FM approved

Compliance

Complies with OSHA 29CFR1910.1030.

Specifications

Certifications/Compliance FM, TUV, OSHA
Diameter (Metric) Outer 35.56 cm
Diameter (English) Outer 14 in.
Capacity (English) 10 gal.
Color White
Height (Metric) 35.5 cm
Includes Foot-operated Cover
For Use With (Application) Safe, temporary collection of biological waste
Height (English) 18 in.
Material Metal
Product Type Biohazard Waste Container
Show More Show Less