missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Biohazard Waste Can

Catalog No. 19033725
Change view
Click to view available options
Capacity (English):
6 gal.
10 gal.
Diameter (English) Outer:
12 in.
14 in.
Height (English):
16 in.
18 in.
Includes:
Foot-operated Cover
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Diameter (English) Outer Height (English) Includes
19033725 6 gal. 12 in. 16 in. Foot-operated Cover
19033755 10 gal. 14 in. 18 in. Foot-operated Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19033725 Supplier Justrite™ Supplier No. 05910
Only null left

The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.

Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.

  • Features leakproof, sturdy steel construction with foot-operated, self-closing lid
  • Eliminates any reason to reach into the waste area or touch the can
  • Holds biohazard bag (sold separately) securely around circumference of can top
  • Large biohazard label clearly identifies the contents and alerts workers

Not to be used for disposal of sharp items.

Certifications

FM approved

Compliance

Complies with OSHA 29CFR1910.1030.

Specifications

Certifications/Compliance FM, TUV, OSHA
Diameter (Metric) Outer 30.2 cm
Diameter (English) Outer 12 in.
Capacity (English) 6 gal.
Color White
Height (Metric) 40.64 cm
Includes Foot-operated Cover
For Use With (Application) Safe, temporary collection of biological waste
Height (English) 16 in.
Material Metal
Product Type Biohazard Waste Container
Show More Show Less