missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Drain Cans

Catalog No. 19018411
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19018411 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19018411 Supplier Justrite™ Supplier No. 11176

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Justrite Drain Cans

Keeps out dirt.

Certifications

FM approved.

Specifications

Certifications/Compliance FM
Color Silver
Quantity 1 Ea.
For Use With (Application) Dry and wet waste collection
For Use With (Equipment) 3 gal. and 5 gal. drain cans
Material Galvanized Steel
Product Type Drain Can Cover