missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Chemcor™ Lined Piggyback Safety Cabinets for Hazardous Materials
GREENER_CHOICE

Catalog No. 19414816
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19414816 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19414816 Supplier Justrite™ Supplier No. 8617282

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To expand storage and safely segregate hazardous materials

  • Used independently or mount on top of standard 114 and 170L (30 and 45 gal.) safety cabinets
  • Non-porous surface resists staining
  • Double-wall, 1mm (18-gauge) welded steel construction
  • Liquid-tight containment sump 51mm (2 in.)
  • Features dual vents, adjustable shelves, three-point stainless steel bullet self-latching doors with U.Loc™ padlockable handle and Haz-Aler™ reflective warning labels
  • Self-close door(s) shuts and latches automatically when a fusible link melts at 74°C (165°F) under fire conditions
  • Includes polyethylene work tray

Certifications

FM approved

Compliance

NFPA 1, NFPA 400 Hazardous Material Code, and the International Fire Code

Warranty

10-Year

Specifications

Product Type ChemCor Lined Piggyback Safety Cabinet
Door Style Self-close
Load Bearing Capacity 17 gal. (64 L)
Width (Metric) Interior 1003 mm
Height (Metric) Interior 495 mm
Height (Metric) Exterior 610 mm
Depth (Metric) Exterior 457 mm
Depth (Metric) Interior 370 mm
Dimensions (D x W x H) Interior 370 x 1003 x 495 mm
Width (Metric) Exterior 1092 mm
No. of Doors 2
Width (English) Exterior 43 in.
Width (English) Interior 39.5 in.
Height (English) Exterior 24 in.
Depth (English) Exterior 18 in.
No. of Shelves 1
Color Blue
Height (English) Interior 19.5 in.
Depth (English) Interior 14.562 in.
Show More Show Less