missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Surface Treated SterileTissue Culture Plates

Catalog No. FB012928
Change view
Click to view available options
Culture Area:
3.85 cm2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
FB012928 3.85 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB012928
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for comparison and analysis of cell growth, yield in multiple platforms.

  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic

Warranty

Three years

Specifications

Array 3 x 4
Certifications/Compliance CE
Bottom Shape Flat
Capacity (Metric) 6.8 mL
Color Natural
Culture Area 3.85 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Standard
Type Cell Culture Plate
Well Count 12
For Use With (Application) Tissue culture
No. per Pack 100 Plates
Packaging Individually wrapped
Show More Show Less