missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Janus Green B

Catalog No. AAA1739114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1739114 25 g
AAA1739106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1739114 Supplier Thermo Scientific Chemicals Supplier No. A1739114
Only null left

CAS: 2869-83-2 | C30H31ClN6 | 511.07 g/mol

Janus Green B is a stain that interacts with DNA and has been used for histology studies including mitochondrial staining, in which its oxidation and reduction reveals alterations to the electron transfer chain. The stain has been utilized in studies for effective and rapid staining of insect chordotonal organs and peripheral nerves, where the nerves appear dark blue in comparison to non stained tissue. With the use of Janus Green B, rabbit lymphatic vessel studies demonstrate that threadlike structures within the lymphatic vessels, known as Bonghan ducts, contain a high density of mitochondria in their cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Janus Green B is a stain that interacts with DNA and has been used for histology studies including mitochondrial staining, in which its oxidation and reduction reveals alterations to the electron transfer chain. The stain has been utilized in studies for effective and rapid staining of insect chordotonal organs and peripheral nerves, where the nerves appear dark blue in comparison to non stained tissue. With the use of Janus Green B, rabbit lymphatic vessel studies demonstrate that threadlike structures within the lymphatic vessels, known as Bonghan ducts, contain a high density of mitochondria in their cells.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol, and slightly soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2869-83-2
Molecular Formula C30H31ClN6
Molecular Weight (g/mol) 511.07
MDL Number MFCD00011758
InChI Key XXACTDWGHQXLGW-UHFFFAOYSA-M
Synonym C.I. 11050
PubChem CID 76123
ChEBI CHEBI:21184
IUPAC Name 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
SMILES [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C

Specifications

Merck Index 14,5255
Solubility Information Soluble in water (20mg/ml at 20°C),ethanol,and slightly soluble in alcohol.
Formula Weight 511.07
Quantity 25 g
Chemical Name or Material Janus Green B
EINECSNumber 220-695-6
RTECSNumber SG1633000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only