missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGA9 Polyclonal Antibody

Description
ITGA9 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.
Spécifications
Spécifications
| Antigène | ITGA9 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gène | Itga9 |
| Numéro d’acquisition du gène | Q13797 |
| Alias du gène | (alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin subunit alpha 9; integrin, alpha 9; ITGA4L; Itga9; Itga9l; RLC |
| Symboles de gène | Itga9 |
| Afficher plus de résultats |