missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IT1t dihydrochloride

Catalog No. 459610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
459610 10 mg
459650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 459610 Supplier Tocris Bioscience™ Supplier No. 4596/10

Potent CXCR4 antagonist

Potent CXCR4 antagonist (IC50 = 1.1 nM in calcium mobilization assays). Orally available. Blocks interaction with the HIV envelope protein, gp120 (IC50 = 7 nM, inhibition of X4-tropic HIV-1IIIB attachment).

Chemical Identifiers

CAS 1092776-63-0
Molecular Formula C21H36Cl2N4S2

Specifications

Quantity 10 mg
Formula Weight 479.57
Purity >99%
Molecular Weight (g/mol) 479.567
Chemical Name or Material IT1t dihydrochloride
Recommended Storage Store at +4°C