accessibility menu, dialog, popup

UserName

H3CNSNSNHNH3CHClHCl(E/Z)

CAS RN 1092776-63-0

IUPAC Name: N,N'-dicyclohexyl[({6,6-dimethyl-5H,6H-imidazo[2,1-b][1,3]thiazol-3-yl}methyl)sulfanyl]methanimidamide dihydrochloride
Synonyms: N,N'-dicyclohexyl({6,6-dimethyl-5H-imidazo[2,1-b][1,3]thiazol-3-yl}methyl)sulfanylmethanimidamide dihydrochloride
Molecular Weight (g/mol): 479.57
Molecular Formula: C21H36Cl2N4S2
InChi Key: HFXJOXOIINQOEB-UHFFFAOYSA-N
SMILES: Cl.Cl.CC1(C)CN2C(CSC(NC3CCCCC3)=NC3CCCCC3)=CSC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results