missing translation for 'onlineSavingsMsg'
Learn More

Isradipine, 98+%

Catalog No. AAJ63920MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63920MF 50 mg
AAJ63920MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63920MF Supplier Thermo Scientific Chemicals Supplier No. J63920MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-type calcium channel blocker | CAS: 75695-93-1 | C19H21N3O5 | 371.39 g/mol

Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

Solubility
Soluble in DMSO

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5
Molecular Weight (g/mol) 371.39
MDL Number MFCD00153820
InChI Key HMJIYCCIJYRONP-UHFFFAOYNA-N
Synonym isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
PubChem CID 3784
IUPAC Name 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C

Specifications

Color Yellow
Quantity 50 mg
Merck Index 14,5240
Solubility Information Soluble in DMSO
Formula Weight 371.39
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isradipine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only