missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole 95.0+%, TCI America™

Catalog No. I03725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I03725G Supplier TCI America Supplier No. I03725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 288-14-2
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00003149
InChI Key CTAPFRYPJLPFDF-UHFFFAOYSA-N
Synonym isoxazole, 1-oxa-2-azacyclopentadiene, isooxazole, unii-00srw0m6pw, 00srw0m6pw, isoxazoles, pubchem8623, acmc-209h4x, ksc204o6n
PubChem CID 9254
ChEBI CHEBI:35595
IUPAC Name 1,2-oxazole
SMILES C1=CON=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 1993
Formula Weight 69.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoxazole