missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carboxylic acid, 98%

Catalog No. AAA1373906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1373906 5 g
AAA1373903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1373906 Supplier Thermo Scientific Chemicals Supplier No. A1373906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21169-71-1 | C4H3NO3 | 113.072 g/mol

Reaction conditions were found to allow the exclusive formation of isoxazole-5-carboxylic acid derivatives by conjugate addition, in acidic medium, of hydroxylamine to β-alkoxyvinyl trichloromethyl ketone in synthesis and reactivity of isoxazoles. Isoxazole-5-carboxylic acid participates in the Synthesis and evaluation of acylguanidine FXa inhibitors. 1, 1, 1-trichloro-4-methoxy-3-penten-2-one postulated in the preparation of isoxazole-5-carboxylic acid from trichloroacetyl chloride, ethyl vinyl ether and hydroxylamine is developed in a fully saturated nitrogen atom theerefore there is no possibility of conjugation with 4- substituents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reaction conditions were found to allow the exclusive formation of isoxazole-5-carboxylic acid derivatives by conjugate addition, in acidic medium, of hydroxylamine to β-alkoxyvinyl trichloromethyl ketone in synthesis and reactivity of isoxazoles. Isoxazole-5-carboxylic acid participates in the Synthesis and evaluation of acylguanidine FXa inhibitors. 1, 1, 1-trichloro-4-methoxy-3-penten-2-one postulated in the preparation of isoxazole-5-carboxylic acid from trichloroacetyl chloride, ethyl vinyl ether and hydroxylamine is developed in a fully saturated nitrogen atom theerefore there is no possibility of conjugation with 4- substituents.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.

Chemical Identifiers

CAS 21169-71-1
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD00156151
InChI Key MIIQJAUWHSUTIT-UHFFFAOYSA-N
Synonym isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
PubChem CID 2060599
IUPAC Name 1,2-oxazole-5-carboxylic acid
SMILES C1=C(ON=C1)C(=O)O

Specifications

Melting Point 141°C to 144°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 113.07
Percent Purity 98%
Chemical Name or Material Isoxazole-5-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only