missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™

Numéro de catalogue. p-2704510
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC10302DA 1g
CC10302EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC10302DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC10302DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 62348-13-4
Formule moléculaire C4H2ClNO2
Poids moléculaire (g/mol) 131.52
Numéro MDL MFCD00067868
Clé InChI NASLINFISOTVJJ-UHFFFAOYSA-N
Synonyme isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, isoxazole-5-carbonylchloride, isoxazole-5-carboxylic acid chloride, acmc-1bhwq, 5-isoxazolecarbonylchloride, isoxazol-5-carbonyl chloride, isoxazole-5-carbonyl-chloride, 5-isoxazolecarbonyl chloride 9ci
PubChem CID 2736707
Nom de l’IUPAC 1,2-oxazole-5-carbonyl chloride
SOURIRES ClC(=O)C1=CC=NO1

Spécifications

Nom ou substance chimique Isoxazole-5-carbonyl chloride
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 131.52
Pourcentage de pureté 90%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.