missing translation for 'onlineSavingsMsg'
Learn More

Lead 207 Isotope Standard for ICP/MS, SPEX CertiPrep™

Catalog No. ISOTPB207
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ISOTPB207 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ISOTPB207 Supplier SPEX CertiPrep Group LLC Supplier No. ISOTPB207

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb 20710μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Lead 207
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Pb
Solvent or Matrix 2% HNO3
Quantity 125 mL
Chemical Name or Material Lead 207 Isotope Standard
Hazard Category Combustible liquid. Suspected of causing cancer. May be fatal if swallowed and enters airways.
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only