missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide 98.0+%, TCI America™

Catalog No. I0407500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0407500G 500 g
I040725G 25 g
I0407100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0407500G Supplier TCI America Supplier No. I0407500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00064827
InChI Key KLDXJTOLSGUMSJ-JGWLITMVSA-N
Synonym isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol
PubChem CID 12597
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES C1C(C2C(O1)C(CO2)O)O

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 500 g
Physical Form Crystal-Powder at 20°C
Boiling Point 175°C
Formula Weight 146.14
Percent Purity ≥98.0% (GC)
Chemical Name or Material Isosorbide
RTECSNumber LZ4380000
TSCA Yes