missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-4-boronic acid 2,2-dimethylpropanediol-1,3 cyclic ester, 97%, Thermo Scientific™

Catalog No. CC04940DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC04940DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC04940DA Supplier Thermo Scientific Chemicals Supplier No. CC04940DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 844891-01-6
Molecular Formula C14H16BNO2
Molecular Weight (g/mol) 241.10
MDL Number MFCD06659058
InChI Key AQEZCUCPPAKCLQ-UHFFFAOYSA-N
Synonym 4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl isoquinoline, isoquinoline-4-boronic acid 2,2-dimethylpropanediol-1,3 cyclic ester, isoquinoline-4-boronic acid, 2,2-dimethylpropane-1,3-diol cyclic ester, isoquinoline-4-boronic acid neopentyl glycol ester, 3 cyclic ester, akos brn-0509, 2-dimethylpropanediol-1, isoquinoline-4-boronic acid 2, 2-4-isoquinolyl-5,5-dimethyl-1,3,2-dioxaborane, isoquinoline,4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl
PubChem CID 2795158
IUPAC Name 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline
SMILES CC1(C)COB(OC1)C1=C2C=CC=CC2=CN=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 241.1
Percent Purity 97%
Chemical Name or Material Isoquinoline-4-boronic acid 2, 2-dimethylpropanediol-1, 3 cyclic ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.