missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoquinoline-1-carbonitrile, 99%
Description
Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 87°C to 91°C |
| Odor | Odorless |
| Quantity | 5 g |
| Beilstein | 119104 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 154.17 |
| Percent Purity | 99% |
| Physical Form | Crystals or Powder or Crystalline Powder or Flakes |
| Chemical Name or Material | Isoquinoline-1-carbonitrile |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H StatementH302-H315-H319-H335 Harmful if swallowed.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only