missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl tetradecanoate, 98%

Catalog No. AAA18111AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA18111AP 500 mL
AAA18111AE 100 mL
AAA181110F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA18111AP Supplier Thermo Scientific Chemicals Supplier No. A18111AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-27-0 | C17H34O2 | 270.457 g/mol

Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

Solubility
Miscible with alcohol. Immiscible with water and glycerol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Density 0.85 g/mL
Boiling Point 191°C to 194°C (20 mmHg)
Flash Point 152°C (306°F)
Assay Percent Range 0.98
Quantity 500 mL
Solubility Information Miscible with alcohol. Immiscible with water and glycerol.
Formula Weight 270.46
Physical Form Liquid
Chemical Name or Material Isopropyl Myristate
EINECSNumber 203-751-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only