missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical
Description
Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 70%, 30% |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| Specific Gravity | 0.872-0.883 |
| UN Number | UN1219 |
| Concentration | 70% |
| Boiling Point | 78.3°C |
| Percent Purity | 68 to 72% |
| Grade | USP |
| Chemical Name or Material | Isopropyl Rubbing Alcohol |