missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Myristate 98.0+%, TCI America™

Catalog No. M0481500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0481500ML 500 mL
M048125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0481500ML Supplier TCI America Supplier No. M0481500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 mL
Formula Weight 270.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Myristate
EINECSNumber (2)-0798
RTECSNumber XB8600000
TSCA Yes