missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl myristate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012451
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012451 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012451 Supplier Merck Emd Millipore Supplier No. PHR11231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isopropyl myristate is an ester of isopropyl alcohol myristic acid. It is mainly used as a solubilizer, emulsifier and emollient in cosmetic and topical medicines. It also finds applications as a flavoring agent in the food industry.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 110-27-0
Density 0.85 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)12 COOCH(CH3)2
MDL Number MFCD00008982
Melting Point ∼3°C
Boiling Point 193°C (20 mmHg)
Synonym IPM; IPM 100; IPM-EX; IPM-R; Isopropyl tetradecanoate; Myristic acid isopropyl ester
Quantity 1 g
Formula Weight 270.45
Grade Certified Reference Material
Show More Show Less