missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyle-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., dioxane cristallin), Fisher BioReagents™
Spécifications
Spécifications
| Nom ou substance chimique | Isopropyl-β-D-thiogalactopyranoside |
| Point de fusion | 122 °C |
| Couleur | Blanc |
| Quantité | 1 g |
| Emballage | Bouteille en verre |
| Forme physique | Solide |
| Indice de Merck | 15, 5126 |
| Poids de formule | 238.3 |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview May form explosive peroxides. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:1 Instability:2 |
| Recommended Storage | -20 °C |