missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Citrate (mixture) 97.0+%, TCI America™

Numéro de catalogue. C103025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C103025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. C103025G Fournisseur TCI America Code fournisseur C103025G
Il en reste null

Identifiants chimiques

CAS 39413-05-3
Molecular Formula C9H14O7
Molecular Weight (g/mol) 234.204
MDL Number MFCD00060079
InChI Key SKHXHUZZFVMERR-UHFFFAOYSA-N
Synonym Citric Acid Isopropyl Ester
PubChem CID 9816081
IUPAC Name 2-hydroxy-2-(2-oxo-2-propan-2-yloxyethyl)butanedioic acid
SMILES CC(C)OC(=O)CC(CC(=O)O)(C(=O)O)O

Spécifications

Color White-Yellow
Quantity 25 g
Percent Purity ≥97.0% (GC)
Physical Form Paste
Chemical Name or Material Isopropyl Citrate (mixture)
RTECSNumber GE7700000
TSCA No