missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Carbamate 98.0+%, TCI America™

Catalog No. C061425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C061425G 25 g
C0614250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C061425G Supplier TCI America Supplier No. C061425G
Only null left

Chemical Identifiers

CAS 1746-77-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00025463
InChI Key OVPLZYJGTGDFNB-UHFFFAOYSA-N
Synonym Carbamic Acid Isopropyl Ester
PubChem CID 15628
IUPAC Name propan-2-yl carbamate
SMILES CC(C)OC(=O)N

Specifications

Melting Point 94°C
Boiling Point 183°C
Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyl Carbamate
EINECSNumber (2)-1712
RTECSNumber FB2975000
TSCA Yes