missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical
I1055, 367-93-1, C9H18O5S
$184.70 - $39,053.24
Chemical Identifiers
| CAS | 367-93-1 |
|---|---|
| Molecular Formula | C9H18O5S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00063273 |
| InChI Key | BPHPUYQFMNQIOC-NXRLNHOXSA-N |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
| SMILES | CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603575
|
N/A | 100 mg | Amber Glass Bottle |
Each for $184.70
|
|
||||
|
18603576
|
N/A | 250 mg | Amber Glass Bottle |
Each for $234.82
|
|
||||
|
18603577
|
N/A | 1 g | Amber Glass Bottle |
Each for $499.06
|
|
||||
|
18603579
|
N/A | 5 g | Amber Glass Bottle |
Each for $1,908.73
|
|
||||
|
18603578
|
N/A | 6 x 1 g | Amber Glass Bottle |
Case of 6 Each for $2,218.03
|
|
||||
|
18603581
|
N/A | 25 g | Amber Glass Bottle |
Each for $7,673.03
|
|
||||
|
18603580
|
N/A | 6 x 5 g | Amber Glass Bottle |
Case of 6 Each for $9,081.70
|
|
||||
|
18603582
|
N/A | 100 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18603583
|
N/A | 1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.Chemical Identifiers
| 367-93-1 | |
| 238.30 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
| C9H18O5S | |
| MFCD00063273 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
Specifications
| 367-93-1 | |
| C9H18O5S | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol | |
| High Purity |
| 100% | |
| MFCD00063273 | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
| 238.30 |