missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical
I1055, 367-93-1, C9H18O5S
Supplier: Spectrum Chemical Mfg Cor I10551GMCS6
| Quantity | 6 x 1 g |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.Chemical Identifiers
| 367-93-1 | |
| 238.30 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
| C9H18O5S | |
| MFCD00063273 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
Specifications
| 367-93-1 | |
| Amber Glass Bottle | |
| MFCD00063273 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
| 100% | |
| C9H18O5S | |
| 6 x 1 g | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
| 238.30 |