missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical

Catalog No. 18603575
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
6 x 1 g
6 x 5 g
100 g
1 kg
Packaging:
Amber Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603575 100 mg Amber Glass Bottle
18603576 250 mg Amber Glass Bottle
18603577 1 g Amber Glass Bottle
18603579 5 g Amber Glass Bottle
18603578 6 x 1 g Amber Glass Bottle
18603581 25 g Amber Glass Bottle
18603580 6 x 5 g Amber Glass Bottle
18603582 100 g Amber Glass Bottle
18603583 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18603575 Supplier Spectrum Chemical Mfg Cor Supplier No. I1055100MG

I1055, 367-93-1, C9H18O5S

Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mg
Grade High Purity
Physical Form Powder or Waxy Solid
Chemical Name or Material Isopropyl-beta-D-thiogalactoside