missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical

I1055, 367-93-1, C9H18O5S

Supplier:  Spectrum Chemical Mfg Cor I10551GM

Quantity 1 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18603577


Only null left

Description

Description

Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Specifications

Chemical Identifiers

367-93-1
238.30
BPHPUYQFMNQIOC-NXRLNHOXSA-N
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
C9H18O5S
MFCD00063273
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol

Specifications

367-93-1
Amber Glass Bottle
MFCD00063273
BPHPUYQFMNQIOC-NXRLNHOXSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
High Purity
100%
C9H18O5S
1 g
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
238.30
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions