missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Technical, Spectrum™ Chemical

Catalog No. 18611132
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611132 500 mL Amber Glass Bottle
18611139 4 L Amber Glass Bottle
18611140 4 L Poly Bottle
18611133 20 L Steel Can
18611131 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611132 Supplier Spectrum Chemical Mfg Cor Supplier No. I1051500MLGL
Only null left

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Boiling Point 78.3°C
Grade Technical
Chemical Name or Material Isopropyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.