missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol Solution, ACS Reagent, 70% Isopropyl Alcohol in Water, Honeywell Burdick & Jackson
$733.92 - $10157.79
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000149
|
Honeywell Inc
LP4204P |
4 x 4 L | Poly Bottle |
Case of 4 Each for $733.92
|
|
||||
|
6000178
|
Honeywell Inc
LP4205G |
5 gal. | Fiberboard Box |
Each for $886.47
|
|
||||
|
6000177
|
Honeywell Chemicals
LP420204P |
204 L | Poly Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Specifications
| 67-63-0 , 7732-18-5 | |
| 70.0000%,30.0000% | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| ACS Reagent | |
| 4 x 4 L | |
| Isopropyl Alcohol Solution, ACS Reagent, 70% Isopropyl Alcohol in Water |
| 87.7°C | |
| C3H8O | |
| 0.785 g/cm3 | |
| Miscible in Water at 20°C | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| Poly Bottle | |
| 11.7°C |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol solution