missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Certified, 70% (v/v), LabChem™

Numéro de catalogue. lc157602
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
LC157602 1 L Poly Bottle
LC157601 500 mL Poly Bottle
LC157604 4 L Poly Bottle
LC157606 10 L Cubitainer
LC157605 20 L Cubitainer
LC15760200 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. LC157602 Fournisseur LabChem Code fournisseur LC157602
Il en reste null

Identifiants chimiques

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Spécifications

Melting Point -88°C
Color Colorless
CAS Max % 70, 30
Decomposition Information Carbon dioxide; Carbon monoxide
Density 0.88g/mL
Quantity 1 L
Flash Point 12°C
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.88g/mL +/- 0.005
Linear Formula C3H8O
Viscosity 2.5316mm2/s
UN Number UN1219
Solubility Information Soluble in water
Boiling Point 82°C
Formula Weight 60.09
Percent Purity 70% (v/v)
Grade Certified
Chemical Name or Material Isopropyl Alcohol
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
EINECSNumber 200-661-7
Recommended Storage Room Temperature