missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >99.9%, Honeywell Burdick & Jackson
Specifications
Specifications
| Absorbance | 0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm |
| CAS Max % | 100.0000% |
| Density | 0.785 g/cm3 |
| Quantity | 56 L |
| Flash Point | 12°C |
| Packaging | Returnable Steel Drum |
| UN Number | UN1219 |
| Water | 0.06% max. |
| Beilstein | 635639 |
| Boiling Point | 82.3°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol |