missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl alcohol, ≥99%, Reagent grade, For ACS analysis, MP Biomedicals™

Catalog No. ICN19400791
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400791 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19400791 Supplier MP Biomedicals Inc Supplier No. 0219400791

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Density 0 .784 to 0.786 g/cm3 at 20°C
Quantity 1 L
Physical Form Liquid
Assay Percent Range ≥99%
Formula Weight 60.1
Grade Reagent
Chemical Name or Material Isopropyl alcohol, For ACS analysis