missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 70%, Spectrum™ Chemical

$174.09 - $6784.06

Chemical Identifiers

CAS 67-63-0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611136
View Documents
Spectrum Chemical Mfg Cor
I1211LTPL
1 L Poly Bottle
Each for $174.09
Only null left
 
18-614-868
View Documents
Spectrum Chemical Mfg Cor
I1214LTPL
4 L Poly Bottle
Each for $294.40
Only null left
 
18611137
View Documents
Spectrum Chemical Mfg Cor
I12120LTCU
20 L Cubitainer
Each for $1,122.74
Only null left
 
18611138
View Documents
Spectrum Chemical Mfg Cor
I121200LTBL
200 L Drum
Each for $6,784.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0

Specifications

67-63-0,7732-18-5
78.3°C
Poly Bottle
70%
Isopropyl Alcohol
70%,30%
1 L
0.877 ± 0.005
Ungraded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.