missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 70%, Spectrum™ Chemical

Catalog No. 18611137
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Cubitainer
Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611137 20 L Cubitainer
18611136 1 L Poly Bottle
18614868 4 L Poly Bottle
18611138 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611137 Supplier Spectrum Chemical Mfg Cor Supplier No. I12120LTCU
Only null left

Chemical Identifiers

CAS 67-63-0 , 7732-18-5

Specifications

CAS Max % 70%, 30%
Boiling Point 78.3°C
Quantity 20 L
Packaging Cubitainer
Specific Gravity 0.877 ± 0.005
Concentration 70%
Grade Ungraded
Chemical Name or Material Isopropyl Alcohol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.