missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 70%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 70%, 30% |
| Boiling Point | 78.3°C |
| Quantity | 1 L |
| Packaging | Poly Bottle |
| Specific Gravity | 0.877 ± 0.005 |
| Concentration | 70% |
| Grade | Ungraded |
| Chemical Name or Material | Isopropyl Alcohol |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.