missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl acetate, 99%

Catalog No. p-7022722
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15618AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA15618AP Supplier Thermo Scientific Chemicals Supplier No. A15618AP

CAS: 108-22-5 | C5H8O2 | 100.12 g/mol

Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

Solubility
Slightly soluble in water. Soluble in alcohol and propylene glycol, acetone and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Density 0.917
Boiling Point 93°C to 94°C
Flash Point 18°C (64°F)
Refractive Index 1.401
Quantity 500 mL
UN Number UN2403
Beilstein 1280347
Merck Index 14,5203
Solubility Information Slightly soluble in water. Soluble in alcohol and propylene glycol,acetone and ethyl ether.
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Isopropenyl acetate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPENYL ACETATE
EINECSNumber 203-562-7
RTECSNumber UD4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only