missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 99+%, spécifié selon les exigences du Ph.Eur.

Numéro de catalogue. AC396982500
Modifier l'affichage
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC396982500 250 mL
AC396980010 1 L
AC396980025 2.5 L
AC396980100 10 L
AC396980250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. AC396982500 Fournisseur Thermo Scientific Chemicals Code fournisseur 396982500

CAS : 67-63-0 | C3H8O | 60,096 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propane-2-ol
SMILES CC(C)O

Spécifications

Melting Point -89.5°C
Absorbance 0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
Color Du brun au violet
Appearance Clear colorless liquid
Density 0.7850g/mL
Quantity 250 mL
Flash Point 12°C
Identification C: Passes Test (EP)
Infrared Spectrum Authentic
Packaging Bouteille en verre
Physical Form Solide de faible fusion
Refractive Index 1.3760 to 1.3780
Specific Gravity 0.785
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa.s (20°C)
Water 0.5% max.
Boiling Point 81.0°C to 83.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Percent Purity ≥99%
Chemical Name or Material Isopropanol
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness
Causes serious eye irritation
Highly flammable liquid and vapor
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray
Wear protective gloves/ protective clothing/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Keep away from heat/sparks/open flames/hot surfaces. - No smoking
Ground/Bond container and receiving equipment

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-661-7

RUO – Usage réservé à la recherche