missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isophthalaldehyde 98.0+%, TCI America™
Supplier: TCI America I0153250G
Specifications
| Isophthalaldehyde | |
| 89°C | |
| 136°C | |
| MFCD00003372 | |
| isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
| C1=CC(=CC(=C1)C=O)C=O | |
| 134.134 | |
| 134.13 | |
| Crystalline Powder |
| 626-19-7 | |
| White-Yellow | |
| C8H6O2 | |
| 250 g | |
| IZALUMVGBVKPJD-UHFFFAOYSA-N | |
| benzene-1,3-dicarbaldehyde | |
| 34777 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 626-19-7 | |
| 134.134 | |
| IZALUMVGBVKPJD-UHFFFAOYSA-N | |
| 34777 | |
| C1=CC(=CC(=C1)C=O)C=O |
| C8H6O2 | |
| MFCD00003372 | |
| isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
| benzene-1,3-dicarbaldehyde |
Safety and Handling
RTECSNumber : NT1981000
TSCA : No