accessibility menu, dialog, popup

UserName

CAS RN 626-19-7

IUPAC Name: benzene-1,3-dicarbaldehyde
Synonyms: 1,3-diformylbenzene
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: IZALUMVGBVKPJD-UHFFFAOYSA-N
SMILES: O=CC1=CC(C=O)=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)