missing translation for 'onlineSavingsMsg'
Learn More

Isophthalaldehyde 98.0+%, TCI America™

Catalog No. p-7131238
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I015325G 25 g
I0153250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I015325G Supplier TCI America Supplier No. I015325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-19-7
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00003372
InChI Key IZALUMVGBVKPJD-UHFFFAOYSA-N
Synonym isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x
PubChem CID 34777
IUPAC Name benzene-1,3-dicarbaldehyde
SMILES C1=CC(=CC(=C1)C=O)C=O

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 134.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalaldehyde
RTECSNumber NT1981000
TSCA No